Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcvrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199964.1 | NP_956258.1 | 1 - 2 | 466 | 98 | |
GenBank mRNA | BC059485.1 | - | 1 - 2 | 466 | 98 | |
KT325592.1 | - | 1 - 2 | 385 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |