Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbp4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199965.1 | NP_956259.1 | 2 - 3 | 185 | 112 | |
GenBank mRNA | BC059516.1 | - | 2 - 3 | 185 | 112 | |
BC152209.1 | - | 2 - 3 | 208 | 112 | ||
BC152210.1 | - | 2 - 3 | 259 | 112 | ||
KM655654.1 | - | 1 - 2 | 122 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |