Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tomm70a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200002.1 | NP_956296.1 | 11 - 12 | 1593 | 106 | |
GenBank mRNA | BC059538.1 | - | 11 - 12 | 1593 | 106 | |
BC171712.1 | - | 11 - 12 | 1693 | 106 | ||
BC171716.1 | - | 11 - 12 | 1693 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |