Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: setd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200054.1 | NP_956348.1 | 6 - 7 | 783 | 70 | |
GenBank mRNA | BC055261.1 | - | 6 - 7 | 783 | 70 | |
BC155278.1 | - | 6 - 7 | 788 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |