Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ciao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200147.2 | NP_956441.2 | 5 - 6 | 795 | 74 | |
GenBank mRNA | BC044534.1 | - | 5 - 6 | 795 | 74 | |
BC065658.1 | - | 5 - 6 | 795 | 74 | ||
BC171538.1 | - | 5 - 6 | 749 | 74 | ||
BC171540.1 | - | 5 - 6 | 749 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |