Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200221.1 | NP_956515.1 | 10 - 11 | 1041 | 93 | |
GenBank mRNA | AY394950.1 | - | 10 - 11 | 1037 | 93 | |
BC046039.1 | - | 10 - 11 | 1041 | 93 | ||
BC152678.1 | - | 10 - 11 | 1121 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |