Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200221.1 | NP_956515.1 | 4 - 5 | 474 | 113 | |
GenBank mRNA | AY394950.1 | - | 4 - 5 | 470 | 113 | |
BC046039.1 | - | 4 - 5 | 474 | 113 | ||
BC152678.1 | - | 4 - 5 | 554 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |