Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200296.1 | NP_956590.1 | 3 - 4 | 205 | 62 | |
XM_005157507.4 | XP_005157564.1 | 3 - 4 | 521 | 62 | ||
XM_005157508.4 | XP_005157565.1 | 3 - 4 | 518 | 62 | ||
XM_005157509.4 | XP_005157566.1 | 3 - 4 | 521 | 62 | ||
XM_005157510.4 | XP_005157567.1 | 3 - 4 | 519 | 62 | ||
XM_009291625.3 | XP_009289900.1 | 3 - 4 | 514 | 62 | ||
GenBank mRNA | BC049481.1 | - | 3 - 4 | 205 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |