Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200296.1 | NP_956590.1 | 8 - 9 | 766 | 101 | |
XM_005157507.4 | XP_005157564.1 | 8 - 9 | 1082 | 101 | ||
XM_005157508.4 | XP_005157565.1 | 8 - 9 | 1049 | 101 | ||
XM_005157509.4 | XP_005157566.1 | 8 - 9 | 1049 | 101 | ||
XM_005157510.4 | XP_005157567.1 | 8 - 9 | 1017 | 101 | ||
XM_009291624.3 | XP_009289899.1 | 8 - 9 | 1076 | 101 | ||
XM_009291625.3 | XP_009289900.1 | 8 - 9 | 1045 | 101 | ||
XM_009291626.3 | XP_009289901.1 | 8 - 9 | 1044 | 101 | ||
XM_021481355.1 | XP_021337030.1 | 8 - 9 | 1071 | 101 | ||
XM_021481356.1 | XP_021337031.1 | 8 - 9 | 1065 | 101 | ||
XM_021481357.1 | XP_021337032.1 | 8 - 9 | 1032 | 101 | ||
GenBank mRNA | BC049481.1 | - | 8 - 9 | 766 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |