Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200466.2 | NP_956760.2 | 8 - 9 | 1158 | 112 | |
GenBank mRNA | BC055169.1 | - | 8 - 9 | 1140 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |