Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stambpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200498.1 | NP_956792.1 | 6 - 7 | 840 | 59 | |
GenBank mRNA | AY398309.1 | - | 6 - 7 | 842 | 59 | |
BC046033.1 | - | 5 - 6 | 636 | 59 | ||
BC055512.1 | - | 6 - 7 | 840 | 59 | ||
BC154224.1 | - | 6 - 7 | 840 | 59 | ||
BC171484.1 | - | 6 - 7 | 752 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |