Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2k18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200512.1 | NP_956806.1 | 1 - 2 | 219 | 75 | |
XM_005163733.4 | XP_005163790.1 | 1 - 2 | 584 | 75 | ||
GenBank mRNA | BC055575.1 | - | 1 - 2 | 219 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |