Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: saga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200559.2 | NP_956853.1 | 3 - 4 | 220 | 66 | |
GenBank mRNA | BC056570.1 | - | 167 | 66 | ||
BC068418.1 | - | 161 | 66 | |||
EB953077.1 | - | 3 - 4 | 205 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |