Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: saga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200559.2 | NP_956853.1 | 4 - 5 | 262 | 89 | |
GenBank mRNA | BC056570.1 | - | 209 | 89 | ||
BC068418.1 | - | 203 | 89 | |||
EB953077.1 | - | 4 - 5 | 247 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |