Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rlbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200705.1 | NP_956999.1 | 6 - 7 | 834 | 97 | |
GenBank mRNA | BC059449.1 | - | 6 - 7 | 834 | 97 | |
EU348853.1 | - | 6 - 7 | 834 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |