Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200726.2 | NP_957020.1 | 5 - 6 | 641 | 62 | |
GenBank mRNA | BC059495.1 | - | 5 - 6 | 616 | 62 |
Gene Symbol: LOC100330746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328713.2 | NP_001315642.1 | 5 - 6 | 651 | 62 | |
NM_001351697.1 | NP_001338626.1 | 5 - 6 | 658 | 62 | ||
GenBank mRNA | BC155195.1 | - | 5 - 6 | 622 | 62 | |
EH449094.1 | - | 5 - 6 | 677 | 62 | ||
EH454751.1 | - | 5 - 6 | 654 | 62 | ||
EH476646.1 | - | 1 - 2 | 323 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |