Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl6aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200734.1 | NP_957028.1 | 7 - 8 | 1881 | 86 | |
XM_017356456.2 | XP_017211945.1 | 7 - 8 | 1913 | 86 | ||
XM_021476813.1 | XP_021332488.1 | 8 - 9 | 2001 | 86 | ||
XM_021476814.1 | XP_021332489.1 | 7 - 8 | 1867 | 86 | ||
XM_021476815.1 | XP_021332490.1 | 7 - 8 | 2176 | 86 | ||
GenBank mRNA | BC059515.1 | - | 7 - 8 | 1881 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |