Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp1r14ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200810.1 | NP_957104.1 | 3 - 4 | 317 | 110 | |
XM_021481361.1 | XP_021337036.1 | 3 - 4 | 191 | 110 | ||
GenBank mRNA | BC059686.1 | - | 3 - 4 | 317 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |