Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kat14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002699.3 | NP_001002699.2 | 7 - 8 | 1465 | 91 | |
GenBank mRNA | CT669750.2 | - | 1 - 2 | 142 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |