Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114628.1 | NP_001108100.1 | 1 - 2 | 649 | 103 | |
XM_005156699.4 | XP_005156756.1 | 2 - 3 | 1156 | 103 | ||
GenBank mRNA | AB292672.1 | - | 1 - 2 | 649 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |