Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsdf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114668.1 | NP_001108140.1 | 2 - 3 | 350 | 69 | |
XM_005173843.4 | XP_005173900.1 | 2 - 3 | 1090 | 69 | ||
XM_009295053.3 | XP_009293328.1 | 2 - 3 | 1090 | 69 | ||
XM_017352819.1 | XP_017208308.1 | 3 - 4 | 427 | 69 | ||
GenBank mRNA | EU378916.1 | - | 2 - 3 | 350 | 69 | |
JF912881.1 | - | 2 - 3 | 318 | 69 | ||
JF912882.1 | - | 2 - 3 | 315 | 69 | ||
JQ182642.1 | - | 2 - 3 | 212 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |