Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hspb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114705.2 | NP_001108177.1 | 1 | 547 | 70 | |
GenBank mRNA | EF583630.1 | - | 1 | 551 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |