Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aqp8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114910.3 | NP_001108382.2 | 1 - 2 | 315 | 66 | |
GenBank mRNA | BC157381.1 | - | 1 - 2 | 284 | 66 | |
EH586667.1 | - | 1 - 2 | 314 | 66 | ||
FJ695516.2 | - | 1 - 2 | 372 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Inventoried Probe spans exons |