Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114925.1 | NP_001108397.1 | 1 - 2 | 492 | 124 | |
XM_005170161.4 | XP_005170218.1 | 2 - 3 | 399 | 124 | ||
GenBank mRNA | BC155622.1 | - | 1 - 2 | 492 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |