Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sobpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114983.2 | NP_001108455.1 | 5 - 6 | 680 | 91 | |
XM_009299931.3 | XP_009298206.1 | 5 - 6 | 987 | 91 | ||
XM_017354921.2 | XP_017210410.1 | 6 - 7 | 2970 | 91 | ||
GenBank mRNA | DQ503413.1 | - | 5 - 6 | 680 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Danio rerio Probe spans exons |