Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115058.1 | NP_001108530.1 | 3 - 4 | 3166 | 109 | |
XM_005159846.4 | XP_005159903.1 | 3 - 4 | 3254 | 109 | ||
GenBank mRNA | EU267178.1 | - | 3 - 4 | 3166 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons |