Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc25b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115095.2 | NP_001108567.1 | 5 - 6 | 739 | 109 | |
GenBank mRNA | EH500454.1 | - | 5 - 6 | 751 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |