Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sept8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115117.1 | NP_001108589.1 | 2 - 3 | 219 | 105 | |
GenBank mRNA | BC055257.1 | - | 2 - 3 | 248 | 105 | |
BC081662.1 | - | 2 - 3 | 219 | 105 | ||
BC100044.1 | - | 2 - 3 | 214 | 105 | ||
BC151876.1 | - | 2 - 3 | 233 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |