Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sept8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115117.1 | NP_001108589.1 | 9 - 10 | 1329 | 60 | |
GenBank mRNA | BC055257.1 | - | 9 - 10 | 1358 | 60 | |
BC081662.1 | - | 9 - 10 | 1329 | 60 | ||
BC100044.1 | - | 9 - 10 | 1324 | 60 | ||
BC151876.1 | - | 9 - 10 | 1343 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |