Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139480.1 | NP_001132952.1 | 4 - 5 | 1069 | 54 | |
GenBank mRNA | AI658050.1 | - | 1 - 2 | 145 | 54 | |
BC129216.1 | - | 4 - 5 | 1038 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |