Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prom1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115143.2 | NP_001108615.2 | 16 - 17 | 2070 | 96 | |
XM_005157347.3 | XP_005157404.1 | 16 - 17 | 2195 | 96 | ||
XM_005157349.3 | XP_005157406.1 | 18 - 19 | 2247 | 96 | ||
XM_005157351.3 | XP_005157408.1 | 17 - 18 | 2223 | 96 | ||
XM_009291267.3 | XP_009289542.1 | 18 - 19 | 2349 | 96 | ||
XM_009291269.3 | XP_009289544.1 | 16 - 17 | 2195 | 96 | ||
XM_021480968.1 | XP_021336643.1 | 18 - 19 | 2350 | 96 | ||
XM_021480969.1 | XP_021336644.1 | 17 - 18 | 2326 | 96 | ||
XM_021480970.1 | XP_021336645.1 | 17 - 18 | 2323 | 96 | ||
XM_021480971.1 | XP_021336646.1 | 18 - 19 | 2349 | 96 | ||
XM_021480972.1 | XP_021336647.1 | 18 - 19 | 2349 | 96 | ||
XM_021480973.1 | XP_021336648.1 | 16 - 17 | 2196 | 96 | ||
XM_021480974.1 | XP_021336649.1 | 18 - 19 | 2350 | 96 | ||
XM_021480975.1 | XP_021336650.1 | 18 - 19 | 2349 | 96 | ||
GenBank mRNA | AF160970.1 | - | 16 - 17 | 2195 | 96 | |
BC163446.1 | - | 16 - 17 | 2070 | 96 | ||
HQ386793.1 | - | 17 - 18 | 2227 | 96 | ||
HQ386794.1 | - | 18 - 19 | 2254 | 96 | ||
HQ386795.1 | - | 16 - 17 | 2195 | 96 | ||
HQ386796.1 | - | 16 - 17 | 2207 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |