Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prom1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115143.2 | NP_001108615.2 | 17 - 18 | 2156 | 81 | |
XM_005157347.3 | XP_005157404.1 | 17 - 18 | 2281 | 81 | ||
XM_005157349.3 | XP_005157406.1 | 19 - 20 | 2333 | 81 | ||
XM_005157351.3 | XP_005157408.1 | 18 - 19 | 2309 | 81 | ||
XM_009291267.3 | XP_009289542.1 | 19 - 20 | 2435 | 81 | ||
XM_009291269.3 | XP_009289544.1 | 17 - 18 | 2281 | 81 | ||
GenBank mRNA | AF160970.1 | - | 17 - 18 | 2281 | 81 | |
BC163446.1 | - | 17 - 18 | 2156 | 81 | ||
HQ386793.1 | - | 18 - 19 | 2313 | 81 | ||
HQ386794.1 | - | 19 - 20 | 2340 | 81 | ||
HQ386795.1 | - | 17 - 18 | 2281 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |