Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prom1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115143.2 | NP_001108615.2 | 21 - 22 | 2448 | 76 | |
XM_005157347.3 | XP_005157404.1 | 21 - 22 | 2573 | 76 | ||
XM_005157349.3 | XP_005157406.1 | 23 - 24 | 2625 | 76 | ||
XM_005157351.3 | XP_005157408.1 | 22 - 23 | 2601 | 76 | ||
XM_009291267.3 | XP_009289542.1 | 23 - 24 | 2727 | 76 | ||
XM_009291269.3 | XP_009289544.1 | 21 - 22 | 2573 | 76 | ||
XM_021480968.1 | XP_021336643.1 | 24 - 25 | 2746 | 76 | ||
XM_021480969.1 | XP_021336644.1 | 23 - 24 | 2722 | 76 | ||
XM_021480970.1 | XP_021336645.1 | 23 - 24 | 2719 | 76 | ||
XM_021480971.1 | XP_021336646.1 | 24 - 25 | 2745 | 76 | ||
XM_021480972.1 | XP_021336647.1 | 24 - 25 | 2745 | 76 | ||
XM_021480973.1 | XP_021336648.1 | 22 - 23 | 2592 | 76 | ||
XM_021480974.1 | XP_021336649.1 | 24 - 25 | 2746 | 76 | ||
XM_021480975.1 | XP_021336650.1 | 24 - 25 | 2745 | 76 | ||
GenBank mRNA | BC163446.1 | - | 21 - 22 | 2448 | 76 | |
HQ386793.1 | - | 22 - 23 | 2605 | 76 | ||
HQ386794.1 | - | 23 - 24 | 2632 | 76 | ||
HQ386795.1 | - | 21 - 22 | 2573 | 76 | ||
HQ386796.1 | - | 22 - 23 | 2603 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |