Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prom1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005157349.3 | XP_005157406.1 | 3 - 4 | 532 | 69 | |
XM_005157351.3 | XP_005157408.1 | 3 - 4 | 532 | 69 | ||
XM_009291267.3 | XP_009289542.1 | 3 - 4 | 634 | 69 | ||
XM_021480968.1 | XP_021336643.1 | 3 - 4 | 635 | 69 | ||
XM_021480969.1 | XP_021336644.1 | 3 - 4 | 635 | 69 | ||
XM_021480971.1 | XP_021336646.1 | 3 - 4 | 634 | 69 | ||
XM_021480972.1 | XP_021336647.1 | 3 - 4 | 634 | 69 | ||
XM_021480974.1 | XP_021336649.1 | 3 - 4 | 635 | 69 | ||
XM_021480975.1 | XP_021336650.1 | 3 - 4 | 634 | 69 | ||
GenBank mRNA | HQ386793.1 | - | 3 - 4 | 536 | 69 | |
HQ386794.1 | - | 3 - 4 | 539 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |