Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: si:ch73-52e5.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122629.1 | NP_001116101.1 | 2 - 3 | 544 | 95 | |
XM_017357412.2 | XP_017212901.1 | 1 - 2 | 825 | 95 | ||
GenBank mRNA | BC159266.1 | - | 2 - 3 | 544 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |