Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122743.1 | NP_001116215.1 | 4 - 5 | 410 | 75 | |
XM_017354919.2 | XP_017210408.1 | 4 - 5 | 450 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |