Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: matr3l1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122784.2 | NP_001116256.2 | 18 - 19 | 3084 | 91 | |
XM_005173122.4 | XP_005173179.1 | 18 - 19 | 3094 | 91 | ||
XM_005173123.4 | XP_005173180.1 | 17 - 18 | 3046 | 91 | ||
XM_009291035.3 | XP_009289310.1 | 17 - 18 | 2193 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |