Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122973.1 | NP_001116445.1 | 3 - 4 | 455 | 73 | |
XM_021478301.1 | XP_021333976.1 | 3 - 4 | 489 | 73 | ||
XM_021478302.1 | XP_021333977.1 | 3 - 4 | 481 | 73 | ||
GenBank mRNA | BC129374.1 | - | 3 - 4 | 624 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |