Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131036.2 | NP_571111.2 | 13 - 14 | 2678 | 85 | |
XM_005171264.4 | XP_005171321.1 | 14 - 15 | 2937 | 85 | ||
XM_005171265.3 | XP_005171322.1 | 12 - 13 | 2409 | 85 | ||
XM_009298633.3 | XP_009296908.1 | 14 - 15 | 2673 | 85 | ||
GenBank mRNA | AJ001423.1 | - | 13 - 14 | 2638 | 85 | |
BC095389.1 | - | 12 - 13 | 2337 | 85 | ||
KM655642.1 | - | 12 - 13 | 2512 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |