Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131036.2 | NP_571111.2 | 6 - 7 | 1465 | 92 | |
XM_005171264.4 | XP_005171321.1 | 6 - 7 | 1460 | 92 | ||
XM_005171265.3 | XP_005171322.1 | 6 - 7 | 1460 | 92 | ||
XM_009298633.3 | XP_009296908.1 | 6 - 7 | 1460 | 92 | ||
GenBank mRNA | AJ001423.1 | - | 6 - 7 | 1425 | 92 | |
BC095389.1 | - | 6 - 7 | 1387 | 92 | ||
KM655642.1 | - | 5 - 6 | 1299 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |