Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200655.3 | NP_956949.3 | 11 - 12 | 1406 | 77 | |
GenBank mRNA | AY398397.1 | - | 11 - 12 | 1407 | 77 | |
BC067626.1 | - | 11 - 12 | 1386 | 77 | ||
BC154301.1 | - | 11 - 12 | 1402 | 77 | ||
EH481640.1 | - | 1 - 2 | 278 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |