Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc37a4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328095.1 | NP_001315024.1 | 9 - 10 | 1965 | 54 | |
XM_005169205.4 | XP_005169262.1 | 9 - 10 | 1511 | 54 | ||
XM_009305596.3 | XP_009303871.1 | 9 - 10 | 1543 | 54 | ||
XM_021479139.1 | XP_021334814.1 | 9 - 10 | 1635 | 54 |
Gene Symbol: LOC108179058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021479503.1 | XP_021335178.1 | 3 - 4 | 221 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Danio rerio Probe spans exons |