Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201017.1 | NP_957311.1 | 2 - 3 | 402 | 69 | |
GenBank mRNA | BC052118.1 | - | 2 - 3 | 402 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |