Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201050.1 | NP_957344.1 | 3 - 4 | 452 | 95 | |
GenBank mRNA | BC048055.1 | - | 3 - 4 | 452 | 95 | |
BC164531.1 | - | 2 - 3 | 350 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |