Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pafah1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201058.1 | NP_957352.1 | 4 - 5 | 389 | 110 | |
GenBank mRNA | BC051609.1 | - | 4 - 5 | 389 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |