Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rorab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201067.1 | NP_957361.1 | 7 - 8 | 1124 | 102 | |
XM_009303216.3 | XP_009301491.1 | 8 - 9 | 1429 | 102 | ||
XM_017356860.2 | XP_017212349.1 | 8 - 9 | 1434 | 102 | ||
XM_021477531.1 | XP_021333206.1 | 7 - 8 | 1399 | 102 | ||
GenBank mRNA | BC051158.1 | - | 7 - 8 | 1124 | 102 | |
BC159203.1 | - | 7 - 8 | 1106 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |