Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201114.1 | NP_957408.1 | 10 - 11 | 1392 | 79 | |
GenBank mRNA | AY330221.1 | - | 10 - 11 | 1372 | 79 | |
BC055237.1 | - | 10 - 11 | 1392 | 79 | ||
BC161664.1 | - | 10 - 11 | 1360 | 79 | ||
BC171389.1 | - | 10 - 11 | 79 | |||
BC171393.1 | - | 10 - 11 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |