Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201114.1 | NP_957408.1 | 7 - 8 | 977 | 96 | |
GenBank mRNA | AY330221.1 | - | 7 - 8 | 957 | 96 | |
BC055237.1 | - | 7 - 8 | 977 | 96 | ||
BC161664.1 | - | 7 - 8 | 945 | 96 | ||
BC171389.1 | - | 7 - 8 | 929 | 96 | ||
BC171393.1 | - | 7 - 8 | 929 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |