Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201114.1 | NP_957408.1 | 9 - 10 | 1175 | 86 | |
GenBank mRNA | AY330221.1 | - | 9 - 10 | 1155 | 86 | |
BC055237.1 | - | 9 - 10 | 1175 | 86 | ||
BC171389.1 | - | 9 - 10 | 1127 | 86 | ||
BC171393.1 | - | 9 - 10 | 1127 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |