Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201118.2 | NP_957412.2 | 6 - 7 | 738 | 75 | |
GenBank mRNA | BC055598.1 | - | 6 - 7 | 611 | 75 | |
BC150464.1 | - | 6 - 7 | 597 | 75 | ||
BC165143.1 | - | 3 - 4 | 227 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |